• USA Home
  • Search Results

  • dimethylterephthalate1941812061611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dimethyl terephthalate

5 Products
Image of Dimethyl terephthalate
CAS Number:
Linear Formula:
 C6H4-1,4-(CO2CH3)2
Molecular Weight:
 194.18
 Beilstein/REAXYS Number:
 1107185
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
 eCl@ss:
 39024601
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Anal. Standard Form
  Shelf Life
  Purity Grade
  Product Line
  Melting Point
  Special Grade
  Application(s)
Show More