• USA Home
  • Search Results

  • dimethylterephthalatesolution1941812061611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dimethyl terephthalate solution

2 Products
Image of Dimethyl terephthalate solution
CAS Number:
Empirical Formula (Hill Notation):
 C10H10O4
Molecular Weight:
 194.18
 MDL number:
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Shelf Life
  Special Grade
  Purity Grade
Show More