• USA Home
  • Search Results

  • dinbutylmagnesiumsolution13853119147511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Di-n-butylmagnesium solution

2 Products
Image of Di-<I>n</I>-butylmagnesium solution
CAS Number:
Linear Formula:
 [CH3(CH2)3]2Mg
Molecular Weight:
 138.53
 Beilstein/REAXYS Number:
 3535184
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Density
Show More