• USA Home
  • Search Results

  • dinotefuran2022116525270011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dinotefuran

2 Products
Synonym:   (RS)-N-Methyl-N′-nitro-N″-[(tetrahydro-3-furanyl)methyl]guanidine
Image of Dinotefuran
CAS Number:
Empirical Formula (Hill Notation):
 C7H14N4O3
Molecular Weight:
 202.21
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Shelf Life
  Special Grade
Show More