• USA Home
  • Search Results

  • dioctylether2424462982311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dioctyl ether

1 Products
Synonym:   Octyl ether
Image of Dioctyl ether
CAS Number:
Linear Formula:
 [CH3(CH2)7]2O
Molecular Weight:
 242.44
 Beilstein/REAXYS Number:
 1748226
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
249599 Dioctyl ether, 99%,