• USA Home
  • Search Results

  • dioctylphthalate3905611781711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dioctyl phthalate

1 Products
Synonym:   Bis(2-ethylhexyl) phthalate, DEHP, DOP, Phthalic acid bis(2-ethylhexyl ester)
Image of Dioctyl phthalate
CAS Number:
Linear Formula:
 C6H4-1,2-[CO2CH2CH(C2H5)(CH2)3CH3]2
Molecular Weight:
 390.56
 Beilstein/REAXYS Number:
 1890696
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More