• USA Home
  • Search Results

  • dioctylsulfide25851269008611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dioctyl sulfide

1 Products
Synonym:   Octyl sulfide
Image of Dioctyl sulfide
CAS Number:
Linear Formula:
 CH3(CH2)7S(CH2)7CH3
Molecular Weight:
 258.51
 Beilstein/REAXYS Number:
 1701624
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
O7007 Dioctyl sulfide, 96%,