• USA Home
  • Search Results

  • diosmetin3002652034311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Diosmetin

3 Products
Synonym:   3′,5,7-Trihydroxy-4′-methoxyflavone, 4′-Methylluteolin, 5,7-Dihydroxy-2-(3-hydroxy-4-methoxyphenyl)chromen-4-one, Luteolin 4′-methyl ether
Image of Diosmetin
CAS Number:
Empirical Formula (Hill Notation):
 C16H12O6
Molecular Weight:
 300.26
 Beilstein/REAXYS Number:
 294492
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
Show More