• USA Home
  • Search Results

  • dipchloridesolution320758511637611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(−)-DIP-Chloride solution

1 Products
Image of (−)-DIP-Chloride<SUP>™</SUP> solution
CAS Number:
Empirical Formula (Hill Notation):
 C20H34BCl
Molecular Weight:
 320.75
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More