• USA Home
  • Search Results

  • dipentene1362313886311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dipentene

2 Products
Synonym:   (±)-Limonene, p-Mentha-1,8-diene
Image of Dipentene
CAS Number:
Empirical Formula (Hill Notation):
 C10H16
Molecular Weight:
 136.23
 Beilstein/REAXYS Number:
 3587825
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Chemical Composition
  Special Grade
Show More