• USA Home
  • Search Results

  • dipentylphthalate3064013118011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dipentyl phthalate

2 Products
Image of Dipentyl phthalate
CAS Number:
Empirical Formula (Hill Notation):
 C18H26O4
Molecular Weight:
 306.40
 Beilstein/REAXYS Number:
 1987323
 EC Number:
 MDL number:
 NACRES:
 NB.61
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Application(s)
  Assay
Show More