• USA Home
  • Search Results

  • diphenylacetylene1782350165511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Diphenylacetylene

1 Products
Synonym:   Tolan
Image of Diphenylacetylene
CAS Number:
Linear Formula:
 C6H5C≡CC6H5
Molecular Weight:
 178.23
 Beilstein/REAXYS Number:
 606478
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
D204803 Diphenylacetylene, 98%,