• USA Home
  • Search Results

  • diphenylamine1692212239411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Diphenylamine

6 Products
Synonym:   DPA
Image of Diphenylamine
CAS Number:
Linear Formula:
 (C6H5)2NH
Molecular Weight:
 169.22
 Beilstein/REAXYS Number:
 508755
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Purity Grade
  Potency
  Melting Point
  Shelf Life
  Application(s)
  Solubility
  Product Line
  Assay
Show More