• USA Home
  • Search Results

  • diphenyleneiodoniumchloride31455467326111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Diphenyleneiodonium chloride

1 Products
Synonym:   DPI, Dibenziodolium chloride
Image of Diphenyleneiodonium chloride
CAS Number:
Empirical Formula (Hill Notation):
 C12H8ClI
Molecular Weight:
 314.55
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More