• USA Home
  • Search Results

  • diphenylphosphorylazide275202638688911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Diphenyl phosphoryl azide

1 Products
Synonym:   DPPA, Phosphoric acid diphenyl ester azide
Image of Diphenyl phosphoryl azide
CAS Number:
Linear Formula:
 (C6H5O)2P(O)N3
Molecular Weight:
 275.20
 Beilstein/REAXYS Number:
 2058967
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More