• USA Home
  • Search Results

  • diphenylsulfide1862713966211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Diphenyl sulfide

3 Products
Synonym:   Phenyl sulfide
Image of Diphenyl sulfide
CAS Number:
Linear Formula:
 (C6H5)2S
Molecular Weight:
 186.27
 Beilstein/REAXYS Number:
 1907932
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Potency
  Titertest
Show More