• USA Home
  • Search Results

  • dipivefrinehydrochloride387906401993811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dipivefrine hydrochloride

2 Products
Synonym:   [2-(2,2-Dimethylpropanoyloxy)-4-[1-hydroxy-2-(methylamino)ethyl]phenyl] 2,2-dimethylpropanoate hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C19H29NO5 · HCl
Molecular Weight:
 387.90
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More