• USA Home
  • Search Results

  • dipropyleneglycolbutylethermixtureofisomers190282991128211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Di(propylene glycol) butyl ether, mixture of isomers

2 Products
Synonym:   DOWANOL® DPnB
Image of Di(propylene glycol) butyl ether, mixture of isomers
CAS Number:
Linear Formula:
 CH3(CH2)3OC3H6OC3H6OH
Molecular Weight:
 190.28
 EC Number:
 NACRES:
 NA.23
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More