• USA Home
  • Search Results

  • directred80137307261010811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Direct Red 80

1 Products
Synonym:   Sirius Red
Image of Direct Red 80
CAS Number:
Empirical Formula (Hill Notation):
 C45H26N10Na6O21S6
Molecular Weight:
 1373.07
 Beilstein/REAXYS Number:
 8185575
 Colour Index Number:
 35780
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More