• USA Home
  • Search Results

  • discoveryrpamidec16hplccolumn1234598765

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Discovery® RP-AmideC16 HPLC Column

11 Products
 NACRES:
 SB.52
 eCl@ss
 32110501
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Composite Dimension
  Operating parameter
  Manufacturer designation
  Matrix active group
  Chromatography Mode
  Application(s)
  Particle Dimension
Show More

Product #
Description
505013 Discovery® RP-AmideC16 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm,
505064 Discovery® RP-AmideC16 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm,
505013-30 Discovery® RP-AmideC16 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 3 mm,
505064-40 Discovery® RP-AmideC16 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4 mm,
569320-U Discovery® RP-AmideC16 HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm,
50501321 Discovery® RP-AmideC16 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm,
505005-30 Discovery® RP-AmideC16 HPLC Column, 5 μm particle size, L × I.D. 5 cm × 3 mm,
505064-30 Discovery® RP-AmideC16 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 3 mm,
569331-U Discovery® RP-AmideC16 HPLC Column, 5 μm particle size, L × I.D. 12.5 cm × 4 mm,
505013-40 Discovery® RP-AmideC16 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4 mm,
505005 Discovery® RP-AmideC16 HPLC Column, 5 μm particle size, L × I.D. 5 cm × 4.6 mm,