• USA Home
  • Search Results

  • disialogangliosidegd1bfrombovinebrain123451955376511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Disialoganglioside GD1b from bovine brain

1 Products
Synonym:   Ganglioside GD1b, disialo
CAS Number:
 MDL number:
 NACRES:
 NA.77
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More