• USA Home
  • Search Results

  • disperseorange131833258169311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Disperse Orange 1

1 Products
Synonym:   4-(4-Nitrophenylazo)diphenylamine
Image of Disperse Orange 1
CAS Number:
Linear Formula:
 O2NC6H4N=NC6H4NHC6H5
Molecular Weight:
 318.33
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Physical Form
Show More