• USA Home
  • Search Results

  • disperseorange25323353148256111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Disperse Orange 25

1 Products
Synonym:   3-[N-Ethyl-4-(4-nitrophenylazo)phenylamino]propionitrile
Image of Disperse Orange 25
CAS Number:
Linear Formula:
 O2NC6H4N=NC6H4N(C2H5)CH2CH2CN
Molecular Weight:
 323.35
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More