• USA Home
  • Search Results

  • disperseorange37392241330161611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Disperse Orange 37

2 Products
Synonym:   3-[4-(2,6-Dichloro-4-nitrophenylazo)-N-ethylanilino]propionitrile
Image of Disperse Orange 37
CAS Number:
Empirical Formula (Hill Notation):
 C17H15Cl2N5O2
Molecular Weight:
 392.24
 Beilstein/REAXYS Number:
 6348868
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Physical Form
  Special Grade
Show More