• USA Home
  • Search Results

  • dispersered1734437317989311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Disperse Red 17

1 Products
Synonym:   3-Methyl-4-(4′-nitrophenylazo)-N,N-bis(2-hydroxyethyl)aniline
Image of Disperse Red 17
CAS Number:
Empirical Formula (Hill Notation):
 C17H20N4O4
Molecular Weight:
 344.37
 Colour Index Number:
 11210
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More