• USA Home
  • Search Results

  • disperseyellow326930283240811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Disperse Yellow 3

2 Products
Synonym:   4-(2-Hydroxy-5-methylphenylazo)acetanilide, N-[4-(2-Hydroxy-5-methylphenylazo)phenyl]acetamide
Image of Disperse Yellow 3
CAS Number:
Empirical Formula (Hill Notation):
 C15H15N3O2
Molecular Weight:
 269.30
 Beilstein/REAXYS Number:
 1915225
 Colour Index Number:
 11855
 EC Number:
 MDL number:
 NACRES:
 NA.47
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Physical Form
Show More