• USA Home
  • Search Results

  • disulfiram296549777811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Disulfiram

4 Products
Synonym:   Bis(diethylthiocarbamoyl) disulfide, Bis(diethylthiocarbamyl) disulfide, Disulfiram, TETD, Tetraethylthiuram disulfide
Image of Disulfiram
CAS Number:
Linear Formula:
 (C2H5)2NCSS2CSN(C2H5)2
Molecular Weight:
 296.54
 Beilstein/REAXYS Number:
 1712560
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Application(s)
  Manufacturer designation
Show More