• USA Home
  • Search Results

  • ditertbutyldicarbonate218252442499511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Di-tert-butyl dicarbonate

4 Products
Synonym:   Di-tert-butyl dicarbonate, Di-tert-butyl pyrocarbonate
Image of Di-tert-butyl dicarbonate
CAS Number:
Empirical Formula (Hill Notation):
 C10H18O5
Molecular Weight:
 218.25
 EC Index Number
 246-240-1
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
Show More