• USA Home
  • Search Results

  • ditertbutyldichlorosilane213221839590911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Di-tert-butyldichlorosilane

1 Products
Synonym:   DTBSCl2
Image of Di-<I>tert</I>-butyldichlorosilane
CAS Number:
Linear Formula:
 [(CH3)3C]2SiCl2
Molecular Weight:
 213.22
 Beilstein/REAXYS Number:
 1739236
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More