• USA Home
  • Search Results

  • diuron2330933054111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Diuron

3 Products
Synonym:   3-(3,4-Dichlorophenyl)-1,1-dimethylurea
Image of Diuron
CAS Number:
Empirical Formula (Hill Notation):
 C9H10Cl2N2O
Molecular Weight:
 233.09
 Beilstein/REAXYS Number:
 2215168
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Shelf Life
  Special Grade
Show More