• USA Home
  • Search Results

  • dl2amino3phosphonopropionicacid169072026306311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

DL-2-Amino-3-phosphonopropionic acid

1 Products
Synonym:   3-Phosphono-DL-alanine, AP-3
Image of <SC>DL</SC>-<WBR>2-<WBR>Amino-<WBR>3-<WBR>phosphonopropionic acid
CAS Number:
Empirical Formula (Hill Notation):
 C3H8NO5P
Molecular Weight:
 169.07
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More