• USA Home
  • Search Results

  • dlactosemonohydrate360316404451511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

D-Lactose monohydrate

4 Products
Synonym:   β-D-Gal-(1→4)-D-Glc, 4-O-β-D-Galactopyranosyl-D-glucose, Milk sugar
Image of <SC>D</SC>-Lactose monohydrate
CAS Number:
Empirical Formula (Hill Notation):
 C12H22O11 · H2O
Molecular Weight:
 360.31
 Beilstein/REAXYS Number:
 3768231
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Restriction
  UV-Absorption Values
  Solubility
  Assay
Show More