• USA Home
  • Search Results

  • dlcycloserine102096839311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

DL-Cycloserine

1 Products
Synonym:   4-Amino-3-isoxazolidinone, 4-Amino-3-isoxazolidone
Image of <SC>DL</SC>-Cycloserine
CAS Number:
Empirical Formula (Hill Notation):
 C3H6N2O2
Molecular Weight:
 102.09
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More