• USA Home
  • Search Results

  • dldithiothreitol15425348312311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

DL-Dithiothreitol

6 Products
Synonym:   threo-1,4-Dimercapto-2,3-butanediol, Cleland’s reagent, DTT
Image of <SC>DL</SC>-Dithiothreitol
CAS Number:
Linear Formula:
 HSCH2CH(OH)CH(OH)CH2SH
Molecular Weight:
 154.25
 Beilstein/REAXYS Number:
 1719757
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  UV-Absorption Condition
  Melting Point
  Absorption
  pH Value
  Special Grade
  UV-Absorption Values
  Product Line
  Assay
Show More