• USA Home
  • Search Results

  • dlisocitricacidtrisodiumsalthydrate25807anhydrousbasis1637736anhydrous11

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

DL-Isocitric acid trisodium salt hydrate

1 Products
Synonym:   threo-DsLs-Isocitric acid trisodium salt hydrate
Image of <SC>DL</SC>-Isocitric acid trisodium salt hydrate
CAS Number:
Empirical Formula (Hill Notation):
 C6H5Na3O7 · xH2O
Molecular Weight:
 258.07 (anhydrous basis)
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More