• USA Home
  • Search Results

  • dlkavain23026315548411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

DL-Kavain

2 Products
Synonym:   trans-5,6-Dihydro-4-methoxy-6-(2-phenylethenyl)-2H-pyran-2-one, DL-Kawain
Image of <SC>DL</SC>-Kavain
CAS Number:
Empirical Formula (Hill Notation):
 C14H14O3
Molecular Weight:
 230.26
 Beilstein Registry Number:
 177877
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Shelf Life
  Special Grade
Show More