• USA Home
  • Search Results

  • dlleucine1311732839211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

DL-Leucine

2 Products
Synonym:   (±)-Amino-4-methylpentanoic acid
Image of <SC>DL</SC>-Leucine
CAS Number:
Linear Formula:
 (CH3)2CHCH2CH(NH2)CO2H
Molecular Weight:
 131.17
 Beilstein/REAXYS Number:
 636005
 EC Number:
 MDL number:
 NACRES:
 NA.26
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Application(s)
  Solubility
  Color
Show More