• USA Home
  • Search Results

  • dlmenthol156278978111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

DL-Menthol

3 Products
Synonym:   2-Isopropyl-5-methylcyclohexanol, Hexahydrothymol, Menthol
Image of <SC>DL</SC>-Menthol
CAS Number:
Empirical Formula (Hill Notation):
 C10H20O
Molecular Weight:
 156.27
 Beilstein/REAXYS Number:
 3194263
 Council of Europe no.:
 63
 EC Number:
 FEMA Number:
 2665
 Flavis number:
 2.015
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Shelf Life
  Optical purity
  Agency/Method
  Assay
Show More