• USA Home
  • Search Results

  • dlmtyrosine1811977506411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

DL-m-Tyrosine

1 Products
Synonym:   (±)-2-Amino-3-(3-hydroxyphenyl)propionic acid, 3-(3-Hydroxyphenyl)-DL-alanine
Image of <SC>DL</SC>-<I>m</I>-Tyrosine
CAS Number:
Linear Formula:
 HOC6H4CH2CH(NH2)CO2H
Molecular Weight:
 181.19
 Beilstein/REAXYS Number:
 2416853
 EC Number:
 MDL number:
 NACRES:
 NA.26
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
T3629 DL-m-Tyrosine, crystalline,