• USA Home
  • Search Results

  • dlpanthenol205251648510211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

DL-Panthenol

1 Products
Synonym:   (±)-α,γ-Dihydroxy-N-(3-hydroxypropyl)-β,β-dimethylbutyramide, (±)-2,4-Dihydroxy-3,3-dimethylbutyric 3-hydroxypropylamide, DL-Pantothenyl alcohol
Image of <SC>DL</SC>-Panthenol
CAS Number:
Linear Formula:
 HOCH2C(CH3)2CH(OH)CONH(CH2)3OH
Molecular Weight:
 205.25
 Beilstein Registry Number:
 1724945
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
295787 DL-Panthenol, 99%,