• USA Home
  • Search Results

  • dlphenylalanine1651915030111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

DL-Phenylalanine

4 Products
Synonym:   (±)-2-Amino-3-phenylpropionic acid
Image of <SC>DL</SC>-Phenylalanine
CAS Number:
Linear Formula:
 C6H5CH2CH(NH2)COOH
Molecular Weight:
 165.19
 Beilstein/REAXYS Number:
 3198807
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
  Product Line
  Melting Point
Show More