• USA Home
  • Search Results

  • dluciferin28032259117511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

D-Luciferin

2 Products
Synonym:   (S)-2-(6-Hydroxy-2-benzothiazolyl)-2-thiazoline-4-carboxylic acid, 4,5-Dihydro-2-(6-hydroxy-2-benzothiazolyl)-4-thiazolecarboxylic acid, Firefly Luciferin
Image of <SC>D</SC>-Luciferin
CAS Number:
Empirical Formula (Hill Notation):
 C11H8N2O3S2
Molecular Weight:
 280.32
 Beilstein Registry Number:
 30484
 EC Number:
 MDL number:
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Biological Source
Show More