• USA Home
  • Search Results

  • dlvaline1171551606311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

DL-Valine

2 Products
Synonym:   (±)-α-Aminoisovaleric acid, DL-2-Amino-3-methylbutanoic acid
Image of <SC>DL</SC>-Valine
CAS Number:
Linear Formula:
 (CH3)2CHCH(NH2)COOH
Molecular Weight:
 117.15
 Beilstein/REAXYS Number:
 506689
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Color
  Special Grade
Show More