• USA Home
  • Search Results

  • dmalicacid1340963661311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

D-Malic acid

1 Products
Synonym:   (R)-(+)-2-Hydroxysuccinic acid, D-(+)-Malic acid, D-Hydroxybutanedioic acid
Image of <SC>D</SC>-Malic acid
CAS Number:
Linear Formula:
 HO2CCH2CH(OH)CO2H
Molecular Weight:
 134.09
 Beilstein/REAXYS Number:
 1723540
 E Number:
 E296
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More