• USA Home
  • Search Results

  • dmannitol182176965811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

D-Mannitol

15 Products
Synonym:   Mannite
Image of <SC>D</SC>-Mannitol
CAS Number:
Empirical Formula (Hill Notation):
 C6H14O6
Molecular Weight:
 182.17
 Beilstein/REAXYS Number:
 1721898
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Restriction
  UV-Absorption Values
  Particle Dimension
  Agency/Method
  Melting Point
  Loss
  Application(s)
  Potency
  Bulk Density
  Autoignition temp
  Special Grade
  pH Value
  Product Line
  Solubility
  Assay
Show More

Product #
Description
M4125 D-Mannitol, ≥98%,
M9647 D-Mannitol, ACS reagent,
M8429 D-Mannitol, meets EP, FCC, USP testing specifications,
M1902 D-Mannitol, suitable for plant cell culture,
M9546 D-Mannitol, BioXtra, ≥98% (HPLC),
63560 D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%,
63559 D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC),
1.37096 D-Mannitol, EMPROVE® EXPERT Ph Eur,BP,USP,JP,
1.05982 D-Mannitol, for microbiology,
78513 D-Mannitol, ≥99.9999% (metals basis), for boron determination,
17311 D-Mannitol, tested according to Ph. Eur.,
1.05980 D-Mannitol, EMPROVE® ESSENTIAL Ph Eur,BP,USP,JP,FCC,E 421,
1.05988 D-Mannitol, fine powder EMPROVE® ESSENTIAL Ph Eur,BP,USP,JP,FCC,E 421,
1.05983 D-Mannitol, for the determination of boric acid,
NIST920 D-Mannitol, NIST® SRM® 920,