• USA Home
  • Search Results

  • dmannoheptulose21018361544911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

D-Mannoheptulose

1 Products
Synonym:   D-Manno-2-heptulose
Image of <SC>D</SC>-Mannoheptulose
CAS Number:
Empirical Formula (Hill Notation):
 C7H14O7
Molecular Weight:
 210.18
 Beilstein/REAXYS Number:
 1726433
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
Show More