• USA Home
  • Search Results

  • dmemnutrientmixturef12ham1234598765

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

DMEM/Nutrient Mixture F-12 Ham

1 Products
Synonym:   DME/F-12, 1:1 mixture, Dulbecco’s Modified Eagle’s Medium/Nutrient Mixture F-12 Ham
 MDL number:
 NACRES:
 NA.75
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More