• USA Home
  • Search Results

  • dmt2fluoroduphosphoramidite7487814695475811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

DMT-2′Fluoro-dU Phosphoramidite

6 Products
Synonym:   DMT-2′-Fluoro-dU amidite
Image of DMT-2′Fluoro-dU Phosphoramidite
CAS Number:
Empirical Formula (Hill Notation):
 C39H46FN4O8P
Molecular Weight:
 748.78
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More