• USA Home
  • Search Results

  • dmt2omethylruphosphoramidite7608111076479911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

DMT-2′O-Methyl-rU Phosphoramidite

5 Products
Synonym:   5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-O-methyl-uridine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite], DMT-2′O-Methyl-rU amidite
Image of DMT-2′O-Methyl-rU Phosphoramidite
CAS Number:
Empirical Formula (Hill Notation):
 C40H49N4O9P
Molecular Weight:
 760.81
 MDL number:
 NACRES:
 NA.51
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More