• USA Home
  • Search Results

  • dmt2otbdmsrcacphosphoramidite9021012105888611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

DMT-2′O-TBDMS-rC(ac) Phosphoramidite

6 Products
Synonym:   N-acetyl-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-O-[(1,1-dimethylethyl)dimethylsilyl]-cytidine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite]
Image of DMT-2′O-TBDMS-rC(ac) Phosphoramidite
CAS Number:
Empirical Formula (Hill Notation):
 C47H64N5O9PSi
Molecular Weight:
 902.10
 MDL number:
 NACRES:
 NA.51
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More